Learn More
Agilent Technologies QUICK CONN CAPILLARY 0.12X220
Catalog No. NC1729686
Encompass
Quick Connect capillary, stainless steel, 0.12 x 220 mm, includes Swagelock fitting
Catalog No. NC1729686 Supplier Agilent Technologies Supplier No. 55001171
Quick Connect capillary, stainless steel, 0.12 x 220 mm, includes Swagelock fitting