Learn More
New Jersey Fluid Systems LLC QUICK CONNECT
Catalog No. NC9763815
Encompass
316 Stainless Steel Swagelok QC4 in. series Quick Connect Shut-OFF Stem; 1/4 in. OD Tube Fitting end; Viton o-ring; 3,000psi @100F
Catalog No. NC9763815 Supplier New Jersey Fluid Systems LLC Supplier No. SSQC4D400
316 Stainless Steel Swagelok QC4 in. series Quick Connect Shut-OFF Stem; 1/4 in. OD Tube Fitting end; Viton o-ring; 3,000psi @100F