Learn More
Neta Scientific QUICK-CONNECT HEAT EXCHNGR ULD
SDP
Catalog No. NC2262481
Encompass_Preferred
This item is manufactured by Agilent
Agilent Quick-Connect Heat Exchanger ULDAgilent Heat exchanger assembly, 1.0-Z, for Agilent 1290 Infinity II LC mercury cadmium telluride (MCT) detector (G7116B) - AA
This item is not returnable. View return policy
Catalog No. NC2262481 Supplier Neta Scientific Supplier No. G711660021
This item is manufactured by Agilent
Agilent Quick-Connect Heat Exchanger ULDAgilent Heat exchanger assembly, 1.0-Z, for Agilent 1290 Infinity II LC mercury cadmium telluride (MCT) detector (G7116B) - AA