Learn More
Analytical Sales & Services Quick Connect Luer Adapter - Female Luer To M6 Female
Catalog No. 5000807070
Encompass
Quick Connect Luer Adapter - Black Female Luer To M6 Female
•Create your own quick connect or adapter
•Low pressure applications
Qty/Pkg: EACH
Catalog No. 50-008-07070 Supplier Analytical Sales & Services Supplier No. 59660
Add to Cart
Add to Cart
Quick Connect Luer Adapter - Black Female Luer To M6 Female
•Create your own quick connect or adapter
•Low pressure applications
Qty/Pkg: EACH