Learn More
Analytical Sales & Services Quick Connect Luer Adapter - Male Luer To M6 Female
Catalog No. 5000805039
Encompass
Quick Connect Luer Adapter - Black Male Luer To M6 Female\n•Create your own quick connect or adapter\n•Low pressure applications\n\nQty/Pkg: EACH
Catalog No. 50-008-05039 Supplier Analytical Sales & Services Supplier No. 59657
Quick Connect Luer Adapter - Black Male Luer To M6 Female\n•Create your own quick connect or adapter\n•Low pressure applications\n\nQty/Pkg: EACH