Learn More
Braintree Scientific Quick Connect Luer Swivels, Dual Luered
SDP
Catalog No. 501955605
Encompass
Quick Connect™ Swivels There is no need to match tubing diameters with swivels any longer. A luer lock on both outlets of dual-luered Quick Connect™ swivel eliminates these complications for good. Generally used with the Quick Connect™ tether the Quick Connect™ swivel is also available with a luer only on one outlet. Use the Quick Connect™ fittings only when needed. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5605 Supplier Braintree Scientific Supplier No. QCSD
Quick Connect™ Swivels There is no need to match tubing diameters with swivels any longer. A luer lock on both outlets of dual-luered Quick Connect™ swivel eliminates these complications for good. Generally used with the Quick Connect™ tether the Quick Connect™ swivel is also available with a luer only on one outlet. Use the Quick Connect™ fittings only when needed. **Sold for research purposes only and not for human use**