Learn More
Grainger Quick-Connect Coupler, 3/8
Catalog No. NC2592579
Encompass_Preferred
The Quick Connect Coupler 3/8" is a push-to-connect fitting made of brass that creates a leak-proof seal for tubing in fluid transfer applications. It has a maximum pressure rating of 232 PSI and temperature range of -40°F to 250°F. This coupler has a 3/8" NPT female thread and is designed for use with 3/8" OD tubing. The fitting provides a quick and easy connect/disconnect method without tools or sealants.
Catalog No. NC2592579 Supplier Grainger Supplier No. 5JKN8
The Quick Connect Coupler 3/8" is a push-to-connect fitting made of brass that creates a leak-proof seal for tubing in fluid transfer applications. It has a maximum pressure rating of 232 PSI and temperature range of -40°F to 250°F. This coupler has a 3/8" NPT female thread and is designed for use with 3/8" OD tubing. The fitting provides a quick and easy connect/disconnect method without tools or sealants.