Learn More
Braintree Scientific Quick Connect Male/Male Spring Tether
SDP
Supplier:  Braintree Scientific QCT18
Encompass
Quick Connect™ Male/Male Spring Tether The Quick Connect™ Spring Tether is intergraded with a polyurethane extension line that is housed within. The tether has male luer ends that connects to the Quick Connect™ Harness and the Quick Connect™ Swivel. Individually packaged and ETO exposed. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5609