Learn More
Grainger Quick-Connect Plug, 3/8 in
Catalog No. NC2592581
Encompass_Preferred
The Quick Connect Plug 3/8 in is a brass, quick connect style plug fitting for use with 3/8 inch tubing. It has a barbed end to insert tubing and a female quick connect end to attach to a male quick connect fitting. This plug is ideal for blocking off quick connect ports or to cap the ends of tubing lines. It is made in the USA out of brass for durability and corrosion resistance. The maximum pressure rating is up to 250 psi. The overall dimensions are 0.5 x 0.5 x 0.75 inches. This is an economical plug for quick connect applications with 3/8 inch tubing.
Catalog No. NC2592581 Supplier Grainger Supplier No. 5JKN4
The Quick Connect Plug 3/8 in is a brass, quick connect style plug fitting for use with 3/8 inch tubing. It has a barbed end to insert tubing and a female quick connect end to attach to a male quick connect fitting. This plug is ideal for blocking off quick connect ports or to cap the ends of tubing lines. It is made in the USA out of brass for durability and corrosion resistance. The maximum pressure rating is up to 250 psi. The overall dimensions are 0.5 x 0.5 x 0.75 inches. This is an economical plug for quick connect applications with 3/8 inch tubing.