Learn More
Grainger EATON HANSEN Hydraulic Quick Connect Hose
Catalog No. NC2585873
Encompass_Preferred
The Eaton Hansen Hydraulic Quick Connect Hose is a high pressure hydraulic hose with quick connect couplings on both ends. It has a working pressure of 5000 psi and a temperature range of -40°F to 212°F. The hose is made of high tensile wire braid reinforcement and has a 1/2" ID. It comes in multiple lengths from 6 inches to 60 inches. The couplings are zinc plated steel with Viton seals. This hydraulic hose is designed for high pressure hydraulic fluid power applications.
Catalog No. NC2585873 Supplier Grainger Supplier No. 2F629
The Eaton Hansen Hydraulic Quick Connect Hose is a high pressure hydraulic hose with quick connect couplings on both ends. It has a working pressure of 5000 psi and a temperature range of -40°F to 212°F. The hose is made of high tensile wire braid reinforcement and has a 1/2" ID. It comes in multiple lengths from 6 inches to 60 inches. The couplings are zinc plated steel with Viton seals. This hydraulic hose is designed for high pressure hydraulic fluid power applications.