Learn More
MSC BluBird BluShield Quick Connect Spray Tip Kit, Brass, 4000 PSI, 5-Piece
Catalog No. 502837057
Encompass
This Quick Connect Spray Tip Kit is designed to enhance pressure washing performance. Made of durable brass, this 5-piece kit is suitable for pressure washers up to 4000 PSI, providing a range of spray patterns for various cleaning applications and includes a dedicated tip for applying cleaning solutions.
  • Replaces old or worn pressure washer spray tips to rejuvenate performance.
  • Includes four high-pressure spray patterns (0, 15, 25, and 40 degrees) for versatile cleaning tasks.
  • Features a dedicated cleaning solution spray tip for enhanced cleaning capabilities for tough grime and stains.
  • Connects to gas pressure washers and quick-connect wands for quick and efficient equipment refresh.
Catalog No. 50-283-7057 Supplier MSC Supplier No. 63950257
This Quick Connect Spray Tip Kit is designed to enhance pressure washing performance. Made of durable brass, this 5-piece kit is suitable for pressure washers up to 4000 PSI, providing a range of spray patterns for various cleaning applications and includes a dedicated tip for applying cleaning solutions.
  • Replaces old or worn pressure washer spray tips to rejuvenate performance.
  • Includes four high-pressure spray patterns (0, 15, 25, and 40 degrees) for versatile cleaning tasks.
  • Features a dedicated cleaning solution spray tip for enhanced cleaning capabilities for tough grime and stains.
  • Connects to gas pressure washers and quick-connect wands for quick and efficient equipment refresh.