Learn More
MSC Value Collection Pneumatic Hose Coupling, 3/8 in Thread, 1 ea
Catalog No. 502867060
Encompass
This pneumatic hose coupling is designed for quick connection applications. It features a 3/8 in thread and a 7/8 in body diameter, constructed from steel with a zinc finish, providing a reliable solution for various industrial needs.
  • Quick connect plug coupling type is featured.
  • Male x female NPT thread type and standard are included.
  • Thread size measures 3/8 in.
  • Body diameter is 7/8 in.
  • Hose inside diameter is 3/8 in.
  • Material is steel with a zinc finish.
  • Country of origin is the United States.
Catalog No. 50-286-7060 Supplier MSC Supplier No. 70057054
This pneumatic hose coupling is designed for quick connection applications. It features a 3/8 in thread and a 7/8 in body diameter, constructed from steel with a zinc finish, providing a reliable solution for various industrial needs.
  • Quick connect plug coupling type is featured.
  • Male x female NPT thread type and standard are included.
  • Thread size measures 3/8 in.
  • Body diameter is 7/8 in.
  • Hose inside diameter is 3/8 in.
  • Material is steel with a zinc finish.
  • Country of origin is the United States.