Learn More
MSC Value Collection Pneumatic Hose Coupling, Quick Connect Socket, 1 ea
Catalog No. 502870139
Encompass
This pneumatic hose coupling is a quick connect socket made from brass, featuring a 1/4 inch thread and a 7/8 inch body diameter.
  • Quick connect socket design
  • Brass construction
  • Thread size: 1/4 inch
  • Body diameter: 7/8 inch
  • Male x female NPT thread type
  • Hose inside diameter: 1/4 inch
Catalog No. 50-287-0139 Supplier MSC Supplier No. 82883463
This pneumatic hose coupling is a quick connect socket made from brass, featuring a 1/4 inch thread and a 7/8 inch body diameter.
  • Quick connect socket design
  • Brass construction
  • Thread size: 1/4 inch
  • Body diameter: 7/8 inch
  • Male x female NPT thread type
  • Hose inside diameter: 1/4 inch