Learn More
MSC Value Collection Quick Connect Socket, Brass, 3/8 NPT, 1 ea
Catalog No. 502866754
Encompass
This brass pneumatic hose coupling functions as a quick connect socket, providing efficient and reliable connections in pneumatic systems. It features a male x female NPT thread type with a 3/8 inch thread size, designed for compatibility with hoses having a 3/8 inch inside diameter.
  • Constructed from durable brass
  • Quick connect socket for rapid assembly
  • Male x female NPT thread configuration
  • Thread size of 3/8 inch
  • Body diameter of 1-1/8 inch
  • Supports 3/8 inch hose inside diameter
Catalog No. 50-286-6754 Supplier MSC Supplier No. 59814616
This brass pneumatic hose coupling functions as a quick connect socket, providing efficient and reliable connections in pneumatic systems. It features a male x female NPT thread type with a 3/8 inch thread size, designed for compatibility with hoses having a 3/8 inch inside diameter.
  • Constructed from durable brass
  • Quick connect socket for rapid assembly
  • Male x female NPT thread configuration
  • Thread size of 3/8 inch
  • Body diameter of 1-1/8 inch
  • Supports 3/8 inch hose inside diameter