Learn More
AVANTOR MASTERFLEX (VWR LLC) Quick-disconnect fittings, Polycarbonate fittings, body, 1/2"" ID, 5/pack
Catalog No. NC1329405
Encompass
CPC (Colder) MPX17803 Quick-Disconnect Body, PC, 1/2" ID; 5/Pk, Sterilization MethodAutoclave, ethylene oxide (ETO), or gamma
Catalog No. NC1329405 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3131113
CPC (Colder) MPX17803 Quick-Disconnect Body, PC, 1/2" ID; 5/Pk, Sterilization MethodAutoclave, ethylene oxide (ETO), or gamma