Learn More
Chemglass Life Sciences Quick Disconnect Female Coupler, 1/2" 316 Stainless Steel, M16 Male End
SDP
Catalog No. 50143646
Encompass_Preferred
Quick Disconnect Female Coupler, 1/2" 316 S.S., M16 Male End. Stainless Steel Quick connects terminating in a male M16 thread. Quick connects have fluorosilicate seals. Both male and female couplers have shutoffs. Quick connects provide a easy and convenient way to change out reactor systems or disconnecting hoses. Temperature range is -60 to 200¿ C. M16 fittings are compatible with the Chemglass ball valves, hoses and adapters.PLEASE NOTE: TWO QUICK CONNECT SETS ARE REQUIRED PER REACTOR SETUP. Special end fittings and configurations are available. CG-1969-Q Quick connects are not considered dripless/dry quick disconnect couplings. Please contact us for details.
Catalog No. 50-143-646 Supplier Chemglass Life Sciences Supplier No. CG1969Q77
Quick Disconnect Female Coupler, 1/2" 316 S.S., M16 Male End. Stainless Steel Quick connects terminating in a male M16 thread. Quick connects have fluorosilicate seals. Both male and female couplers have shutoffs. Quick connects provide a easy and convenient way to change out reactor systems or disconnecting hoses. Temperature range is -60 to 200¿ C. M16 fittings are compatible with the Chemglass ball valves, hoses and adapters.PLEASE NOTE: TWO QUICK CONNECT SETS ARE REQUIRED PER REACTOR SETUP. Special end fittings and configurations are available. CG-1969-Q Quick connects are not considered dripless/dry quick disconnect couplings. Please contact us for details.