Learn More
MSC BluShield Quick Connect Spray Tip, 40° Spray Angle, Brass
Catalog No. 502837007
Encompass
This quick connect spray tip ensures precision and versatility in pressure washing. Its 40° spray angle, identified by its white color, is ideal for delicate surfaces like painted finishes, protecting integrity while delivering optimal results. It connects directly to pressure washer wands with a 1/4 in quick disconnect socket.
  • Unparalleled precision and versatility.
  • Engineered for specific spray pattern needs.
  • Quick disconnect design for adaptability.
  • 40° spray angle (white color) for delicate surfaces.
  • Ensures optimal results and preserves surface integrity.
  • Connects to 1/4 in quick disconnect pressure washer wands.
  • Offers flexibility for selecting the perfect spray pattern.
  • Enhances precision and efficiency.
Catalog No. 50-283-7007 Supplier MSC Supplier No. 63956346
This quick connect spray tip ensures precision and versatility in pressure washing. Its 40° spray angle, identified by its white color, is ideal for delicate surfaces like painted finishes, protecting integrity while delivering optimal results. It connects directly to pressure washer wands with a 1/4 in quick disconnect socket.
  • Unparalleled precision and versatility.
  • Engineered for specific spray pattern needs.
  • Quick disconnect design for adaptability.
  • 40° spray angle (white color) for delicate surfaces.
  • Ensures optimal results and preserves surface integrity.
  • Connects to 1/4 in quick disconnect pressure washer wands.
  • Offers flexibility for selecting the perfect spray pattern.
  • Enhances precision and efficiency.