Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MCD2404 Quick-disconnect fittings, Metal NPT (M) pipe adapters, inserts, valved, 1/4"" NPT
Catalog No. NC1452580
Encompass
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.
Catalog No. NC1452580 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3140461
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.