Learn More
Perkin Elmer US LLC Quick Disconnect Tubing without Swagelock
Catalog No. 502103735
Encompass_Preferred
This tubing is included in the Cyclonic/Concentric Torch Module for Optima 8x00 (N0780607) and the Cyclonic/Concentric Torch Module for Optima 2x00/4x00/5x00/7x00 (N0770607). The Quick Disconnect Tubing with Swagelock can be purchased separately (N0770336).
Catalog No. 50-210-3735 Supplier Perkin Elmer US LLC Supplier No. N0770610
This tubing is included in the Cyclonic/Concentric Torch Module for Optima 8x00 (N0780607) and the Cyclonic/Concentric Torch Module for Optima 2x00/4x00/5x00/7x00 (N0770607). The Quick Disconnect Tubing with Swagelock can be purchased separately (N0770336).