Learn More
Research Products International Corp Quick-DNA Fecal/Soil Microbe Miniprep Kit, 50 Preps
SDP
Catalog No. 501977410
Encompass
The Quick-DNA Fecal/Soil Microbe Miniprep Kit is a fecal and soil DNA extraction kit designed for the simple and rapid isolation of inhibitor-free, PCR-quality host cell and microbial DNA from a variety of sample sources, including humans, birds, rats, mice, cattle, etc. The procedure is easy and can be completed in minutes: fecal samples are rapidly and efficiently lysed by bead beating with our state of the art, ultra-high density BashingBeads. Zymo-Spin column technology is then used to isolate the DNA, which is subsequently filtered to remove humic acids/polyphenols that can inhibit PCR. Eluted DNA is ideal for downstream molecular-based applications including PCR, arrays, genotyping, methylation detection, etc.
Rapid method for the isolation of inhibitor-free, PCR-quality DNA from fecal samples in minutes, including from humans, birds, rats, mice, cattle, etc.
Catalog No. 50-197-7410 Supplier Research Products International Corp Supplier No. ZD6010
The Quick-DNA Fecal/Soil Microbe Miniprep Kit is a fecal and soil DNA extraction kit designed for the simple and rapid isolation of inhibitor-free, PCR-quality host cell and microbial DNA from a variety of sample sources, including humans, birds, rats, mice, cattle, etc. The procedure is easy and can be completed in minutes: fecal samples are rapidly and efficiently lysed by bead beating with our state of the art, ultra-high density BashingBeads. Zymo-Spin column technology is then used to isolate the DNA, which is subsequently filtered to remove humic acids/polyphenols that can inhibit PCR. Eluted DNA is ideal for downstream molecular-based applications including PCR, arrays, genotyping, methylation detection, etc.
Rapid method for the isolation of inhibitor-free, PCR-quality DNA from fecal samples in minutes, including from humans, birds, rats, mice, cattle, etc.