Learn More
Zymo Research Corporation Quick-DNA HMW MagBead Kit (96 prep)
SDP
Catalog No. 501957614
Encompass_Preferred
Innovative reagents and Zymo Researchs unique system allow for a simple Bind, Wash, & Elute procedure that is unmatched in providing ultra-pure and high yield DNA of > 150 kb size. Isolated DNA is ready for immediate use in sensitive downstream applications including long read (Oxford Nanopore™) and NGS sequencing, qPCR, arrays, and methylation analysis.
Catalog No. 50-195-7614 Supplier Zymo Research Corporation Supplier No. D6060
Innovative reagents and Zymo Researchs unique system allow for a simple Bind, Wash, & Elute procedure that is unmatched in providing ultra-pure and high yield DNA of > 150 kb size. Isolated DNA is ready for immediate use in sensitive downstream applications including long read (Oxford Nanopore™) and NGS sequencing, qPCR, arrays, and methylation analysis.