Learn More
Zymo Research Corporation Quick-DNA™ Plus 96 Magbead Kit (1 x 96 Preps)
SDP
Catalog No. NC1669398
Encompass_Preferred
The Quick-DNA™ MagBead Plus Kit is the innovative reagents and Zymo Researchs unique system allow for a simple Bind, Wash, & Elute procedure that is unmatched in providing ultrapure and high yielding DNA (up to 150 kb). Isolated DNA is ready for immediate use in sensitive downstream applications including DNA sequencing (NGS and long read), qPCR, arrays, and methylation analysis.
Catalog No. NC1669398 Supplier Zymo Research Corporation Supplier No. D4081
The Quick-DNA™ MagBead Plus Kit is the innovative reagents and Zymo Researchs unique system allow for a simple Bind, Wash, & Elute procedure that is unmatched in providing ultrapure and high yielding DNA (up to 150 kb). Isolated DNA is ready for immediate use in sensitive downstream applications including DNA sequencing (NGS and long read), qPCR, arrays, and methylation analysis.