Learn More
Zymo Research Corporation Quick DNA Universal Microprep Kit (50 Preps)
SDP
Catalog No. 501251437
Encompass_Preferred
For high yield total DNA extraction (e.g., genomic, mitochondrial, viral), cell culture, solid tissue, saliva, and any biological fluid sample. Innovative reagents and Zymo-Spin Technology allow for ultra-pure and concentrated genomic DNA > 50 kb to be eluted in as little as 10 l. Zymo-Spin Columns ensure no buffer retention. Purified DNA is RNA-free bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays, and methylation analysis.
Catalog No. 50-125-1437 Supplier Zymo Research Corporation Supplier No. D4074
For high yield total DNA extraction (e.g., genomic, mitochondrial, viral), cell culture, solid tissue, saliva, and any biological fluid sample. Innovative reagents and Zymo-Spin Technology allow for ultra-pure and concentrated genomic DNA > 50 kb to be eluted in as little as 10 l. Zymo-Spin Columns ensure no buffer retention. Purified DNA is RNA-free bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays, and methylation analysis.