Learn More
Zymo Research Corporation Quick-DNA™ Midiprep Plus Kit (25 Preps)
SDP
Catalog No. 501251438
Encompass_Preferred
The Quick-DNA™ Midiprep Plus Kit facilitates rapid and efficient purification of DNA from any biological fluids, cultured/monolayer cells, and solid tissues by combining enzymatic and chemical extraction regimens. Innovative reagents and Zymo-Spin™Technology allow for ultra-pure and concentrated genomic DNA 50 kb to be eluted in as little as 200 l. Zymo-Spin™ Columns ensure no buffer retention. Purified DNA is RNA-free, bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays, and methylation analysis.
Catalog No. 50-125-1438 Supplier Zymo Research Corporation Supplier No. D4075
The Quick-DNA™ Midiprep Plus Kit facilitates rapid and efficient purification of DNA from any biological fluids, cultured/monolayer cells, and solid tissues by combining enzymatic and chemical extraction regimens. Innovative reagents and Zymo-Spin™Technology allow for ultra-pure and concentrated genomic DNA 50 kb to be eluted in as little as 200 l. Zymo-Spin™ Columns ensure no buffer retention. Purified DNA is RNA-free, bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays, and methylation analysis.