Learn More
Zymo Research Corporation Quick-DNA™ Miniprep Plus Kit (10 Preps)
SDP
Catalog No. 501251425
Encompass_Preferred
The Quick-DNA™ Miniprep Plus Kit is the easiest method for high yield total DNA extraction (e.g., genomic, plasmid, mitochondrial, viral) from any biological fluid,cell culture, or solid tissue sample. Innovative reagents and Zymo-Spin™ Technology allow for ultra-pure and concentrated genomic DNA 50 kb to be eluted in as little as 35 l. Zymo-Spin™ Columns ensure no buffer retention. Purified DNA is RNA-free, bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays, and methylation analysis.
Catalog No. 50-125-1425 Supplier Zymo Research Corporation Supplier No. D4068T
The Quick-DNA™ Miniprep Plus Kit is the easiest method for high yield total DNA extraction (e.g., genomic, plasmid, mitochondrial, viral) from any biological fluid,cell culture, or solid tissue sample. Innovative reagents and Zymo-Spin™ Technology allow for ultra-pure and concentrated genomic DNA 50 kb to be eluted in as little as 35 l. Zymo-Spin™ Columns ensure no buffer retention. Purified DNA is RNA-free, bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays, and methylation analysis.