Learn More
Research Products International Corp Quick-DNA Miniprep Plus Kit, 200 Preps
SDP
Catalog No. 501977111
Encompass
The Quick-DNA Plus Kits are the easiest DNA isolation kits for high yield total DNA extraction (e.g., genomic, mitochondrial, viral) from cell culture, solid tissue, saliva, and any biological fluid sample. Innovative reagents and Zymo-Spin Technology allow for ultra-pure and concentrated genomic DNA (> 50 kb) to be eluted in as little as 10 µl. Zymo-Spin Columns ensure no buffer retention. Purified DNA is RNA-free,removing the need for RNase A treatments and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays, and methylation analysis.
Quick & Easy: Simple 20 minute procedure.
Highest Yield: Recover 3x more DNA.
Ultra-Pure: Ready for qPCR, Next-Gen Sequencing, arrays, etc.
Catalog No. 50-197-7111 Supplier Research Products International Corp Supplier No. ZD4069
The Quick-DNA Plus Kits are the easiest DNA isolation kits for high yield total DNA extraction (e.g., genomic, mitochondrial, viral) from cell culture, solid tissue, saliva, and any biological fluid sample. Innovative reagents and Zymo-Spin Technology allow for ultra-pure and concentrated genomic DNA (> 50 kb) to be eluted in as little as 10 µl. Zymo-Spin Columns ensure no buffer retention. Purified DNA is RNA-free,removing the need for RNase A treatments and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays, and methylation analysis.
Quick & Easy: Simple 20 minute procedure.
Highest Yield: Recover 3x more DNA.
Ultra-Pure: Ready for qPCR, Next-Gen Sequencing, arrays, etc.