Learn More
Research Products International Corp Quick-DNA Plant/Seed Miniprep Kit, 50 Preps
SDP
Catalog No. 501977412
Encompass
The Quick-DNA Plant/Seed Miniprep Kit is a plant DNA isolation kit designed for the simple, rapid isolation of inhibitor-free, PCR-quality DNA from a variety of plant sample sources including leaves, stems, buds, flowers, fruit, seeds, etc. The procedure is easy and can be completed in minutes: plant samples are rapidly and efficiently lysed by bead beating with our state of the art, ultra-high density BashingBeads. Polysaccharides, lipids, and polyphenols/tannins are removed from the DNA using our Zymo-Spin column technology with Zymo-Spin IV-IR HRC filter. The eluted DNA is ideal for downstream molecular-based applications including PCR, arrays, etc.
Rapid and simple method for DNA isolation from tough-to-lyse plant and seed samples.
Ultra-high density BashingBeads are fracture resistant and chemically inert.
Zymo-Spin column technology coupled with filtration removes polyphenolic PCR inhibitors from the DNA product.
Catalog No. 50-197-7412 Supplier Research Products International Corp Supplier No. ZD6020
The Quick-DNA Plant/Seed Miniprep Kit is a plant DNA isolation kit designed for the simple, rapid isolation of inhibitor-free, PCR-quality DNA from a variety of plant sample sources including leaves, stems, buds, flowers, fruit, seeds, etc. The procedure is easy and can be completed in minutes: plant samples are rapidly and efficiently lysed by bead beating with our state of the art, ultra-high density BashingBeads. Polysaccharides, lipids, and polyphenols/tannins are removed from the DNA using our Zymo-Spin column technology with Zymo-Spin IV-IR HRC filter. The eluted DNA is ideal for downstream molecular-based applications including PCR, arrays, etc.
Rapid and simple method for DNA isolation from tough-to-lyse plant and seed samples.
Ultra-high density BashingBeads are fracture resistant and chemically inert.
Zymo-Spin column technology coupled with filtration removes polyphenolic PCR inhibitors from the DNA product.