Learn More
Zymo Research Corporation Quick-DNA™ Plus 96 Magbead Kit (4 x 96 Preps)
SDP
Catalog No. 502037075
Encompass_Preferred
The Quick-DNA™ MagBead Plus Kit is the innovative reagents and Zymo Researchs unique system allow for a simple Bind, Wash, & Elute procedure that is unmatched in providing ultrapure and high yielding DNA (up to 150 kb). Isolated DNA is ready for immediate use in sensitive downstream applications including DNA sequencing (NGS and long read), qPCR, arrays, and methylation analysis.
Catalog No. 50-203-7075 Supplier Zymo Research Corporation Supplier No. D4082
The Quick-DNA™ MagBead Plus Kit is the innovative reagents and Zymo Researchs unique system allow for a simple Bind, Wash, & Elute procedure that is unmatched in providing ultrapure and high yielding DNA (up to 150 kb). Isolated DNA is ready for immediate use in sensitive downstream applications including DNA sequencing (NGS and long read), qPCR, arrays, and methylation analysis.