Learn More
Zymo Research Corporation Quick-DNA/RNA FFPE Kit (50 Preps)
SDP
Catalog No. 501251664
Encompass_Preferred
The unique chemistries of the product have been optimized for maximum recovery of DNA and of both large and small RNA species. Simply deparaffinize tissues using Deparaffinization Solution, digest using Proteinase K, heat to reverse chemical crosslinks, and then purify using Zymo-Spin™ Column technology. The result is high-quality DNA & total RNA (including small RNAs) that is DNA-free and is ready for RT/qPCR, hybridization, sequencing, etc.
Catalog No. 50-125-1664 Supplier Zymo Research Corporation Supplier No. R1009
The unique chemistries of the product have been optimized for maximum recovery of DNA and of both large and small RNA species. Simply deparaffinize tissues using Deparaffinization Solution, digest using Proteinase K, heat to reverse chemical crosslinks, and then purify using Zymo-Spin™ Column technology. The result is high-quality DNA & total RNA (including small RNAs) that is DNA-free and is ready for RT/qPCR, hybridization, sequencing, etc.