Learn More
Zymo Research Corporation Quick-DNA/RNA HT
SDP
Catalog No. 502865902
Encompass_Preferred
TheQuick-DNA/RNAHTkit is intended for rapid, high-throughput nucleic acid extraction (automated or manual) from any biological or clinical sample (e.g., swabs – nasal/nasopharyngeal, oropharyngeal, etc.; biological liquids – blood, plasma, serum, saliva, sputum, cells in suspension, etc.; tissue – needle biopsies, LCM, etc.) and/or samples stored in most collection matrices and devices (e.g., UTM, VTM, *DNA/RNA Shield™, RNAlater, RNAProtect, etc.). The kit is compatible with robotic-type sample processors (i.e, bead movers) in combination with sensitive downstream molecular amplification assays. High-quality DNA/RNA extracted with theQuick-DNA/RNAHTkit can be used for Next-Gen sequencing, RT/qPCR and more.
Catalog No. 50-286-5902 Supplier Zymo Research Corporation Supplier No. R2150
TheQuick-DNA/RNAHTkit is intended for rapid, high-throughput nucleic acid extraction (automated or manual) from any biological or clinical sample (e.g., swabs – nasal/nasopharyngeal, oropharyngeal, etc.; biological liquids – blood, plasma, serum, saliva, sputum, cells in suspension, etc.; tissue – needle biopsies, LCM, etc.) and/or samples stored in most collection matrices and devices (e.g., UTM, VTM, *DNA/RNA Shield™, RNAlater, RNAProtect, etc.). The kit is compatible with robotic-type sample processors (i.e, bead movers) in combination with sensitive downstream molecular amplification assays. High-quality DNA/RNA extracted with theQuick-DNA/RNAHTkit can be used for Next-Gen sequencing, RT/qPCR and more.