Learn More
Zymo Research Corporation Quick-DNA/RNA™ MagBead (96 preps)
SDP
Catalog No. NC1848865
Encompass_Preferred
The extraction method has the option to recover total nucleic acids in one elution or DNA & RNA in separate elutions without the use of phenol.DNA/RNA is eluted into 50 l of DNase/RNase-Free Water and is ready for any downstream application including Next-Gen Sequencing, RT/PCR, hybridization, etc.
Catalog No. NC1848865 Supplier Zymo Research Corporation Supplier No. R2130
The extraction method has the option to recover total nucleic acids in one elution or DNA & RNA in separate elutions without the use of phenol.DNA/RNA is eluted into 50 l of DNase/RNase-Free Water and is ready for any downstream application including Next-Gen Sequencing, RT/PCR, hybridization, etc.