Learn More
Zymo Research Corporation Quick-DNA/RNA Microprep Plus Kit (10 Preps)
SDP
Catalog No. 501957597
Encompass_Preferred
The procedure uses unique spin-column technology that results in highquality DNA and total RNA (including small RNAs 17-200 nt) and is ready for Next-Gen Sequencing, RT/qPCR, hybridization, etc.
Catalog No. 50-195-7597 Supplier Zymo Research Corporation Supplier No. D7005T
The procedure uses unique spin-column technology that results in highquality DNA and total RNA (including small RNAs 17-200 nt) and is ready for Next-Gen Sequencing, RT/qPCR, hybridization, etc.