Learn More
Research Products International Corp Quick-DNA/RNA MiniPrep Kit, 50 Preps
SDP
Catalog No. 501977248
Encompass
The Quick-DNA/RNA Miniprep Kit provides a quick method for the isolation of high quality genomic DNA and total RNA from small amounts of cells and tissue. The kit isolates both genomic DNA and a broad range of RNA species without the use of phenol. Small RNAs (e.g., tRNAs, microRNAs) can be recovered following a simple adjustment within the RNA isolation protocol – no extra steps are required! Both DNA and RNA from up to 5x106 cells can be eluted into volumes as little as 25 µl in less than 15 minutes.
Quick & Easy: Extract DNA and RNA from cells, small amounts of easy-to-lyse tissue, buffy coat, buccal cells, plasma, serum, and other biological liquids. Not compatible with whole blood.
Ultra-Pure: Ready for Next-generation sequencing, RT-qPCR, arrays, etc. DNase I included (Plus kit and MagBead only)
Catalog No. 50-197-7248 Supplier Research Products International Corp Supplier No. ZD7001
The Quick-DNA/RNA Miniprep Kit provides a quick method for the isolation of high quality genomic DNA and total RNA from small amounts of cells and tissue. The kit isolates both genomic DNA and a broad range of RNA species without the use of phenol. Small RNAs (e.g., tRNAs, microRNAs) can be recovered following a simple adjustment within the RNA isolation protocol – no extra steps are required! Both DNA and RNA from up to 5x106 cells can be eluted into volumes as little as 25 µl in less than 15 minutes.
Quick & Easy: Extract DNA and RNA from cells, small amounts of easy-to-lyse tissue, buffy coat, buccal cells, plasma, serum, and other biological liquids. Not compatible with whole blood.
Ultra-Pure: Ready for Next-generation sequencing, RT-qPCR, arrays, etc. DNase I included (Plus kit and MagBead only)