Learn More
Zymo Research Corporation Quick DNA/RNA MiniPrep Plus Kit (10 Preps.)
SDP
Catalog No. 502037181
Encompass_Preferred
The procedure uses unique spin-column technology that results in highquality DNA and total RNA (including small RNAs 17-200 nt) and is ready for Next-Gen Sequencing, RT/qPCR, hybridization, etc.
Catalog No. 50-203-7181 Supplier Zymo Research Corporation Supplier No. D7003T
The procedure uses unique spin-column technology that results in highquality DNA and total RNA (including small RNAs 17-200 nt) and is ready for Next-Gen Sequencing, RT/qPCR, hybridization, etc.