Learn More
Research Products International Corp Quick-DNA/RNA MiniPrep Plus Kit, 50 Preps
SDP
Catalog No. 501977181
Encompass
The Quick-DNA/RNA MiniPrep Plus kit combines Quick-DNA/RNA technology with the addition of DNA/RNA Shield, a unique preservation and lysis technology, and Proteinase K to enable easy, reliable, and rapid isolation from any biological sample including cells, solid tissue, and whole blood. The procedure uses Zymo-Spin column technology that results in high-quality genomic DNA and total RNA that is ready for any downstream application including reverse transcription, microarray, sequencing, etc.
Quick & Easy: Extract DNA and RNA from any sample including cells, solid tissue, whole blood, biological liquids, FFPE tissue, environmental (plant/seed), swabs (stool, soil, microbial samples), etc.
Ultra-Pure: Ready for Next-generation sequencing, RT-qPCR, arrays, etc. DNase I included (Plus kit and MagBead only)
Catalog No. 50-197-7181 Supplier Research Products International Corp Supplier No. ZD7003
The Quick-DNA/RNA MiniPrep Plus kit combines Quick-DNA/RNA technology with the addition of DNA/RNA Shield, a unique preservation and lysis technology, and Proteinase K to enable easy, reliable, and rapid isolation from any biological sample including cells, solid tissue, and whole blood. The procedure uses Zymo-Spin column technology that results in high-quality genomic DNA and total RNA that is ready for any downstream application including reverse transcription, microarray, sequencing, etc.
Quick & Easy: Extract DNA and RNA from any sample including cells, solid tissue, whole blood, biological liquids, FFPE tissue, environmental (plant/seed), swabs (stool, soil, microbial samples), etc.
Ultra-Pure: Ready for Next-generation sequencing, RT-qPCR, arrays, etc. DNase I included (Plus kit and MagBead only)