Learn More
Zymo Research Corporation Quick-DNA/RNA Pathogen MagBead (96 preps)
SDP
Catalog No. 502037270
Encompass_Preferred
The kit features a storage/lysis buffer system and can be combined with high density ZR BashingBead™ Lysis Tubes (*recommended) to facilitate complete homogenization of hard-to-lyse samples for efficient nucleic acid isolation. Small (>50 nt) and large (> 200 kb) DNA and RNA are bound to magnetic beads, washed and then eluted.The isolated high-quality nucleic acids are suitable for all downstream applications such as Next-Gen sequencing, hybridization-based and RT/qPCR detection.
Catalog No. 50-203-7270 Supplier Zymo Research Corporation Supplier No. R2145
The kit features a storage/lysis buffer system and can be combined with high density ZR BashingBead™ Lysis Tubes (*recommended) to facilitate complete homogenization of hard-to-lyse samples for efficient nucleic acid isolation. Small (>50 nt) and large (> 200 kb) DNA and RNA are bound to magnetic beads, washed and then eluted.The isolated high-quality nucleic acids are suitable for all downstream applications such as Next-Gen sequencing, hybridization-based and RT/qPCR detection.