Learn More
Zymo Research Corporation Quick-DNA/RNA™ Pathogen MiniPrep (50 Preps.)
SDP
Catalog No. 502037242
Encompass_Preferred
The kit features a storage/lysis buffer system and can be combined with high density ZR BashingBead™ Lysis Tubes (*recommended) to facilitate complete homogenization of hard-to-lyse samples for efficient nucleic acid isolation. Small (>50 nt) and large (> 200 kb) DNA and RNA are bound to the column, washed and then eluted. The isolated high-quality nucleic acids are suitable for all downstream applications such as Next-Gen sequencing, hybridization-based and RT/qPCR detection.
Catalog No. 50-203-7242 Supplier Zymo Research Corporation Supplier No. R1042
The kit features a storage/lysis buffer system and can be combined with high density ZR BashingBead™ Lysis Tubes (*recommended) to facilitate complete homogenization of hard-to-lyse samples for efficient nucleic acid isolation. Small (>50 nt) and large (> 200 kb) DNA and RNA are bound to the column, washed and then eluted. The isolated high-quality nucleic acids are suitable for all downstream applications such as Next-Gen sequencing, hybridization-based and RT/qPCR detection.