Learn More
Zymo Research Corporation Quick-DNA/RNA Viral Kit (2x96)
SDP
Catalog No. 502037185
Encompass_Preferred
The kit also features a buffer system that facilitates complete viral particle lysis for efficient nucleic acid isolation. Small (> 50 nt) and large (> 200 kb) DNA and RNA are bound to each well of the plate, washed and eluted.The isolated high-quality nucleic acids are ready for all downstream applications such as Next-Gen sequencing, hybridization-based and RT/qPCR detection.
Catalog No. 50-203-7185 Supplier Zymo Research Corporation Supplier No. D7022
The kit also features a buffer system that facilitates complete viral particle lysis for efficient nucleic acid isolation. Small (> 50 nt) and large (> 200 kb) DNA and RNA are bound to each well of the plate, washed and eluted.The isolated high-quality nucleic acids are ready for all downstream applications such as Next-Gen sequencing, hybridization-based and RT/qPCR detection.