Learn More
Zymo Research Corporation Quick-DNA Tissue/Insect 96 Kit (2 x 96 Preps)
SDP
Catalog No. 502037175
Encompass_Preferred
The procedure is easy and can be completed in as little as 40 minutes. Samples are added directly to the tubes of a ZR BashingBead™ Lysis Rack (2.0 mm) and rapidly and efficiently lysed by bead beating (e.g., 2010 GenoGrinder Instrument, page 4) without using organic denaturants or proteinases. The DNA is isolated and purified using our Zymo-Spin™ Technology and is ideal for downstream molecular-based applications including PCR, array, genotyping, etc. PCR inhibitors are effectively removed during the purification process.
Catalog No. 50-203-7175 Supplier Zymo Research Corporation Supplier No. D6017
The procedure is easy and can be completed in as little as 40 minutes. Samples are added directly to the tubes of a ZR BashingBead™ Lysis Rack (2.0 mm) and rapidly and efficiently lysed by bead beating (e.g., 2010 GenoGrinder Instrument, page 4) without using organic denaturants or proteinases. The DNA is isolated and purified using our Zymo-Spin™ Technology and is ideal for downstream molecular-based applications including PCR, array, genotyping, etc. PCR inhibitors are effectively removed during the purification process.