Learn More
Zymo Research Corporation Quick-DNA™ Viral 96 Kit (4 x 96 Preps)
SDP
Catalog No. 501251363
Encompass_Preferred
Quick recovery of viral DNA from a wide range of sources using Zymo-Spin plate technology. Plate design allows DNA to be eluted at high concentrations into minimal volumes. Eluted DNA is suitable for PCR, Southern blotting, and restriction endonuclease digestion.
Catalog No. 50-125-1363 Supplier Zymo Research Corporation Supplier No. D3018
Quick recovery of viral DNA from a wide range of sources using Zymo-Spin plate technology. Plate design allows DNA to be eluted at high concentrations into minimal volumes. Eluted DNA is suitable for PCR, Southern blotting, and restriction endonuclease digestion.