Learn More
Zymo Research Corporation Quick-DNA™ Viral 96 Kit (2 x 96 Preps)
SDP
Catalog No. 501957622
Encompass_Preferred
Quick recovery of viral DNA from a wide range of sources using Zymo-Spin plate technology. Plate design allows DNA to be eluted at high concentrations into minimal volumes. Eluted DNA is suitable for PCR, Southern blotting, and restriction endonuclease digestion.
Catalog No. 50-195-7622 Supplier Zymo Research Corporation Supplier No. D3017
Quick recovery of viral DNA from a wide range of sources using Zymo-Spin plate technology. Plate design allows DNA to be eluted at high concentrations into minimal volumes. Eluted DNA is suitable for PCR, Southern blotting, and restriction endonuclease digestion.