Learn More
Zymo Research Corporation Quick-DNA™ Viral Kit (200 Preps) w/ Zymo-Spin™ IC Columns (Capped)
SDP
Catalog No. 501251361
Encompass_Preferred
Quick recovery of viral DNA from a wide range of sources using Zymo-Spin column technology. Column design allows DNA to be eluted at high concentrations into minimal volumes. Eluted DNA is suitable for PCR, Southern blotting, and restriction endonuclease digestion.
Catalog No. 50-125-1361 Supplier Zymo Research Corporation Supplier No. D3016
Quick recovery of viral DNA from a wide range of sources using Zymo-Spin column technology. Column design allows DNA to be eluted at high concentrations into minimal volumes. Eluted DNA is suitable for PCR, Southern blotting, and restriction endonuclease digestion.