Learn More
COLDER PRODUCTS COMPANY CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Body Polysulfone Diameter Inner 6 4 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber
Catalog No. 503131304
Encompass_Preferred
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Body Polysulfone Diameter Inner 6 4 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber
Catalog No. 50-313-1304 Supplier COLDER PRODUCTS COMPANY Supplier No. MPC17004T39
CPC Plastic Quick-Disconnect Fittings Hose Barb Non-Valved In-line Coupling Body Polysulfone Diameter Inner 6 4 mm Autoclavable Autoclavable Certifications/Compliance USP Class VI ADCF Amber