Learn More
Grainger Quick Exhaust Valve 1/8 In NPT
Catalog No. 502750933
Encompass_Preferred
The Pneumadyne Quick Exhaust Valve is a 1/8" NPT brass body valve that allows rapid exhaust of compressed air to atmosphere. It has a maximum pressure rating of 125 psi and a temperature range of -20 to 180°F. This normally closed valve features a chrome-plated brass ball and stem with Buna-N seals. When actuated, it exhausts downstream air in less than 0.5 seconds. The valve body measures 1-1/4" in length. This quick exhaust valve enables rapid depressurization in pneumatic applications.
Catalog No. 50-275-0933 Supplier Grainger Supplier No. 14F303
The Pneumadyne Quick Exhaust Valve is a 1/8" NPT brass body valve that allows rapid exhaust of compressed air to atmosphere. It has a maximum pressure rating of 125 psi and a temperature range of -20 to 180°F. This normally closed valve features a chrome-plated brass ball and stem with Buna-N seals. When actuated, it exhausts downstream air in less than 0.5 seconds. The valve body measures 1-1/4" in length. This quick exhaust valve enables rapid depressurization in pneumatic applications.