Learn More
LGC GENOMICS LLC (Lucigen) QuickExtract DNA Extraction Solution 1.0, 50 mL
Catalog No. NC9904870
Encompass
The QuickExtract™ DNA Extraction Solution can be used to rapidly and efficiently extract PCR-ready genomic DNA from almost any sample type using a simple, one-tube protocol that takes only 3-8 minutes, depending on the sample.
This item is not returnable. View return policy
Catalog No. NC9904870 Supplier LGC GENOMICS LLC Supplier No. QE09050
The QuickExtract™ DNA Extraction Solution can be used to rapidly and efficiently extract PCR-ready genomic DNA from almost any sample type using a simple, one-tube protocol that takes only 3-8 minutes, depending on the sample.