Learn More
Zymo Research Corporation Quick-DNA Fecal/Soil Microbe MidiPrep™ Kit (25 Preps)
SDP
Catalog No. 501251611
Encompass_Preferred
The Quick-DNA Fecal/Soil Microbe Midiprep Kit is designed for the simple and rapid isolation of inhibitor-free, PCR-quality host cell and microbial DNA from a variety of sample sources, including humans, birds, rats, mice, cattle, etc. The procedure is easy and can be completed in minutes: fecal samples are rapidly and efficiently lysed by bead beating with our state of the art, ultra-high density BashingBeads. Zymo-Spin column technology is then used to isolate the DNA which is subsequently filtered to remove humic acids/polyphenols that can inhibit PCR. Eluted DNA is ideal for downstream molecular-based applications including PCR, arrays, genotyping, methylation detection, etc.
Catalog No. 50-125-1611 Supplier Zymo Research Corporation Supplier No. D6110
The Quick-DNA Fecal/Soil Microbe Midiprep Kit is designed for the simple and rapid isolation of inhibitor-free, PCR-quality host cell and microbial DNA from a variety of sample sources, including humans, birds, rats, mice, cattle, etc. The procedure is easy and can be completed in minutes: fecal samples are rapidly and efficiently lysed by bead beating with our state of the art, ultra-high density BashingBeads. Zymo-Spin column technology is then used to isolate the DNA which is subsequently filtered to remove humic acids/polyphenols that can inhibit PCR. Eluted DNA is ideal for downstream molecular-based applications including PCR, arrays, genotyping, methylation detection, etc.