Learn More
Electron Microscopy Sciences Quick Jig Mounting Fixture
SDP
Supplier:  Electron Microscopy Sciences 5016005
Encompass
Versatile spring-loaded fixture for securing samples with pressure during mounting using adhesives, waxes, and epoxies. The fixture can be used at room temp or on a hot plate for heat curable epoxies or faster drying of conductive paint. A scale on the pressure rod allows the user to apply reproducible pressure. Two interchangeable trays accommodate six different sized disk sample mounts ( 1/2", 3/4", 1", 1 1/4", 1 1/2", and 2" diameters) and five angled SEM pin stubs (0 deg, 45 deg, 52 deg, 70 deg, and 90 deg). The angled SEM pin stubs are held with screws on the angled surface so that the sample lies horizontally and the pressure applied vertically normal to the sample's surface. The 70 degree angle is useful for pretilted EBSD sample holders and the 52 degree angle is useful for FIB holders.The "PTFE" tip contacting the sample is compliant and easily cleaned if any adhesive inadvertently comes in contact with it. The fixture adapts to a large range in sample height.
Catalog No. 50-192-6830