Learn More
New England Biolabs, Inc. Quick-Load 100 bp DNA Ladder – 1.25 ml
SDP
Catalog No. 50994942
Encompass
Quick-Load 100 bp DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing bromophenol blue as a tracking dye. The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 12 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100-1,517 base pairs. The 500 and 1,000 base pair bands have increased intensity to serve as reference bands. NEB also offers a Quick-Load version of this ladder with purple dye.
Catalog No. 50-994-942 Supplier New England Biolabs, Inc. Supplier No. N0467S
Quick-Load 100 bp DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing bromophenol blue as a tracking dye. The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 12 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100-1,517 base pairs. The 500 and 1,000 base pair bands have increased intensity to serve as reference bands. NEB also offers a Quick-Load version of this ladder with purple dye.