Learn More
New England Biolabs, Inc. Quick-Load 1 kb Plus DNA Ladder – 1.25 ml
SDP
Catalog No. 50994896
Encompass
Quick-Load 1 kb Plus DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing bromophenol blue as a tracking dye. The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis. This digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference bands. NEB also offers a Quick-Load version of this ladder with purple dye.
Catalog No. 50-994-896 Supplier New England Biolabs, Inc. Supplier No. N0469S
Quick-Load 1 kb Plus DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing bromophenol blue as a tracking dye. The DNA Ladder consists of proprietary plasmids which are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis. This digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference bands. NEB also offers a Quick-Load version of this ladder with purple dye.