Learn More
New England Biolabs, Inc. Quick-Load Taq 2X Master Mix – 500 reactions (50 µl vol)
SDP
Catalog No. 50995171
Encompass
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5' to 3' polymerase activity and a 5' flap endonuclease activity. Quick-Load Taq 2X Master Mix is an optimized ready-to-use solution containing Taq DNA Polymerase, dNTPs, MgCl2, KCl, tracking dyes and stabilizers. The presence of two commonly used tracking dyes for DNA gels, Orange G and Xylene Cyanol FF, gives the master mix a green color, allowing direct loading of the PCR product onto agarose gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at approximately 4 kb and Orange G migrates at approximately 50 bp. The amount of tracking dyes included does not mask co-migrating DNA bands. Quick-Load Taq 2X Master Mix is ideally suited to routine PCR applications from templates including pure DNA solutions, bacterial colonies, and cDNA products. It can amplify up to 4 kb from complex genomic DNA or up to 5 kb from lambda DNA.
Catalog No. 50-995-171 Supplier New England Biolabs, Inc. Supplier No. M0271L
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5' to 3' polymerase activity and a 5' flap endonuclease activity. Quick-Load Taq 2X Master Mix is an optimized ready-to-use solution containing Taq DNA Polymerase, dNTPs, MgCl2, KCl, tracking dyes and stabilizers. The presence of two commonly used tracking dyes for DNA gels, Orange G and Xylene Cyanol FF, gives the master mix a green color, allowing direct loading of the PCR product onto agarose gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at approximately 4 kb and Orange G migrates at approximately 50 bp. The amount of tracking dyes included does not mask co-migrating DNA bands. Quick-Load Taq 2X Master Mix is ideally suited to routine PCR applications from templates including pure DNA solutions, bacterial colonies, and cDNA products. It can amplify up to 4 kb from complex genomic DNA or up to 5 kb from lambda DNA.